siRNA Details
| Gene Symbol: Usp43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291049.1 | NP_001277978.1 | 2 | 651 | ||
| NM_173754.4 | NP_776115.2 | 2 | 651 | |||
| GenBank mRNA | AK083271.1 | - | 2 | 489 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp43| Gene Symbol: Usp43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291049.1 | NP_001277978.1 | 2 | 651 | ||
| NM_173754.4 | NP_776115.2 | 2 | 651 | |||
| GenBank mRNA | AK083271.1 | - | 2 | 489 | ||