siRNA Details
| Gene Symbol: Ube2o | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173755.3 | NP_776116.2 | 15, 16 | 2970 | ||
| GenBank mRNA | AK045398.1 | - | 2, 3 | 340 | ||
| AK129431.1 | - | 15, 16 | 2970 | |||
| BC023329.1 | - | 2, 3 | 700 | |||
| BC039233.1 | - | 7, 8 | 1436 | |||
| BC051108.1 | - | 6, 7 | 1315 | |||
| BC055067.1 | - | 1, 2 | 63 | |||
| BC059193.1 | - | 15, 16 | 2595 | 4160667 | ||

Ube2o
Genomic Map