siRNA Details
| Gene Symbol: Rab6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173781.4 | NP_776142.1 | 2 | 186 | ||
| GenBank mRNA | AB232597.1 | - | 2 | 103 | ||
| AK035893.1 | - | 2 | 145 | |||
| AK082732.1 | - | 2 | 309 | |||
| BC060618.1 | - | 2 | 186 | 6406134 | ||

Rab6b
Genomic Map