siRNA Details
| Gene Symbol: Disc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174853.2 | NP_777278.2 | 2 | 161 | ||
| NM_174854.2 | NP_777279.2 | 2 | 161 | |||
| GenBank mRNA | AF513723.1 | - | 2 | 161 | ||
| AF513724.1 | - | 2 | 161 | |||
| AJ506179.1 | - | 2 | 126 | |||
| AJ506180.1 | - | 2 | 147 | |||
| AY177673.1 | - | 2 | 126 | |||
| AY320287.1 | - | 3 | 243 | |||

Disc1
Genomic Map