siRNA Details
| Gene Symbol: Eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175009.3 | NP_778174.1 | 5 | 1032 | ||
| GenBank mRNA | AK016562.1 | - | 5 | 2417 | ||
| AK032957.1 | - | 5 | 1032 | |||
| AK136334.1 | - | 1 | 119 | |||
| AK162511.1 | - | 5 | 1027 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eny2| Gene Symbol: Eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175009.3 | NP_778174.1 | 5 | 1032 | ||
| GenBank mRNA | AK016562.1 | - | 5 | 2417 | ||
| AK032957.1 | - | 5 | 1032 | |||
| AK136334.1 | - | 1 | 119 | |||
| AK162511.1 | - | 5 | 1027 | |||