siRNA Details
| Gene Symbol: Aqp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175087.4 | NP_780296.1 | 1 | 380 | ||
| GenBank mRNA | AK082699.1 | - | 1 | 381 | ||
| BC115586.1 | - | 1 | 381 | 40102369 | ||
| BC115587.1 | - | 1 | 380 | |||
| DQ464060.1 | - | 1 | 381 | |||
| DQ826418.1 | - | 1 | 372 | |||

Aqp6
Genomic Map