siRNA Details
| Gene Symbol: Sf3b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175102.4 | NP_780311.2 | 1 | 650 | ||
| GenBank mRNA | AK003475.1 | - | 1 | 619 | ||
| AK009874.1 | - | 1 | 613 | |||
| AK010348.1 | - | 1 | 625 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sf3b5| Gene Symbol: Sf3b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175102.4 | NP_780311.2 | 1 | 650 | ||
| GenBank mRNA | AK003475.1 | - | 1 | 619 | ||
| AK009874.1 | - | 1 | 613 | |||
| AK010348.1 | - | 1 | 625 | |||