siRNA Details
| Gene Symbol: Hspbap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175111.3 | NP_780320.2 | 1, 2 | 162 | ||
| GenBank mRNA | AA879889.1 | - | 1, 2 | 164 | ||
| AK017731.1 | - | 1, 2 | 128 | |||
| AK028824.1 | - | 1, 2 | 143 | |||
| AK049404.1 | - | 1, 2 | 140 | |||
| AK076189.1 | - | 1, 2 | 129 | |||
| AK089941.1 | - | 1, 2 | 177 | |||
| AK138906.1 | - | 1, 2 | 142 | |||

Hspbap1
Genomic Map