siRNA Details
| Gene Symbol: Fbxo28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175127.2 | NP_780336.1 | 2 | 357 | ||
| GenBank mRNA | AK076211.1 | - | 2 | 363 | ||
| AK077653.1 | - | 2 | 370 | |||
| AK122295.1 | - | 2 | 342 | |||
| AK162091.1 | - | 2 | 326 | |||
| AK162188.1 | - | 2 | 238 | |||
| BC055458.1 | - | 1 | 59 | |||
| BC094542.1 | - | 2 | 357 | 6408744 | ||

Fbxo28
Genomic Map