siRNA Details
| Gene Symbol: Ube2o | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173755.3 | NP_776116.2 | 17 | 3194 | ||
| GenBank mRNA | AK035970.1 | - | 1 | 326 | ||
| AK045398.1 | - | 4 | 564 | |||
| AK129431.1 | - | 17 | 3194 | |||
| BC023329.1 | - | 4 | 924 | |||
| BC039233.1 | - | 9 | 1660 | |||
| BC051108.1 | - | 8 | 1539 | |||
| BC055067.1 | - | 3 | 286 | |||
| BC059193.1 | - | 17 | 2819 | 4160667 | ||

Ube2o
Genomic Map