siRNA Details
| Gene Symbol: SLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304743.1 | NP_001291672.1 | 1, 2 | 719 | ||
| NM_014720.3 | NP_055535.2 | 1, 2 | 719 | |||
| XM_011540401.2 | XP_011538703.1 | 1, 2 | 731 | |||
| GenBank mRNA | AB002804.1 | - | 1, 2 | 390 | ||
| AF273048.1 | - | 1, 2 | 192 | |||
| BC047762.1 | - | 1, 2 | 712 | |||
| BC047885.1 | - | 1, 2 | 719 | |||
| BC064804.1 | - | 1, 2 | 715 | |||
| BC111565.1 | - | 1, 2 | 189 | 40009070 | ||
| D86959.1 | - | 1, 2 | 669 | |||

SLK
Genomic Map