siRNA Details
| Gene Symbol: Mrps27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173757.3 | NP_776118.2 | 2, 3 | 163 | ||
| GenBank mRNA | AK076008.1 | - | 2, 3 | 162 | ||
| BC049149.1 | - | 2, 3 | 91 | 6309524 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps27| Gene Symbol: Mrps27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173757.3 | NP_776118.2 | 2, 3 | 163 | ||
| GenBank mRNA | AK076008.1 | - | 2, 3 | 162 | ||
| BC049149.1 | - | 2, 3 | 91 | 6309524 | ||