siRNA Details
| Gene Symbol: 4933406M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173771.4 | NP_776132.2 | 4 | 679 | ||
| GenBank mRNA | AK030161.1 | - | 4 | 681 | ||
| BC116694.1 | - | 3 | 352 | 40105550 | ||
| BC116697.1 | - | 3 | 352 | 40105553 | ||

4933406M09Rik
Genomic Map