siRNA Details
| Gene Symbol: Oog4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173773.1 | NP_776134.1 | 3 | 474 | ||
| GenBank mRNA | AK054346.1 | - | 3 | 496 | ||
| AY573564.1 | - | 3 | 474 | |||
| BC053722.1 | - | 2 | 345 | |||
| BC098485.1 | - | 2 | 333 | 30051767 | ||

Oog4
Genomic Map