siRNA Details
| Gene Symbol: Gimap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174960.2 | NP_777620.1 | 2 | 965 | ||
| GenBank mRNA | AV291572.2 | - | 1 | 213 | ||
| BC096680.1 | - | 2 | 949 | 1345477 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gimap9| Gene Symbol: Gimap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174960.2 | NP_777620.1 | 2 | 965 | ||
| GenBank mRNA | AV291572.2 | - | 1 | 213 | ||
| BC096680.1 | - | 2 | 949 | 1345477 | ||