siRNA Details
| Gene Symbol: Hpcal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174998.4 | NP_778163.1 | 5 | 851 | ||
| GenBank mRNA | AK032591.1 | - | 5 | 851 | ||
| BC138631.1 | - | 4 | 686 | 8861651 | ||
| BC138632.1 | - | 4 | 686 | 8861652 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hpcal4| Gene Symbol: Hpcal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174998.4 | NP_778163.1 | 5 | 851 | ||
| GenBank mRNA | AK032591.1 | - | 5 | 851 | ||
| BC138631.1 | - | 4 | 686 | 8861651 | ||
| BC138632.1 | - | 4 | 686 | 8861652 | ||