siRNA Details
| Gene Symbol: Hspbap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175111.3 | NP_780320.2 | 2, 3 | 342 | ||
| GenBank mRNA | AA879889.1 | - | 2, 3 | 344 | ||
| AK017731.1 | - | 2, 3 | 308 | |||
| AK028824.1 | - | 2, 3 | 323 | |||
| AK049404.1 | - | 2, 3 | 320 | |||
| AK076189.1 | - | 2, 3 | 309 | |||
| AK089941.1 | - | 2, 3 | 357 | |||
| AK138906.1 | - | 2, 3 | 322 | |||

Hspbap1
Genomic Map