siRNA Details
| Gene Symbol: Trak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175114.3 | NP_780323.2 | 3, 4 | 511 | ||
| GenBank mRNA | AK029123.1 | - | 4, 5 | 909 | ||
| AK041436.1 | - | 4, 5 | 879 | |||
| AK052264.1 | - | 4, 5 | 898 | |||
| AK122426.1 | - | 4, 5 | 601 | |||
| BC046285.1 | - | 3, 4 | 501 | |||
| BC058971.1 | - | 3, 4 | 453 | 6832507 | ||
| BC116262.1 | - | 3, 4 | 478 | |||
| BC116263.1 | - | 3, 4 | 478 | |||
| BC129961.1 | - | 3, 4 | 478 | |||
| BC129962.1 | - | 3, 4 | 478 | |||

Trak1
Genomic Map