siRNA Details
| Gene Symbol: Fbxo28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175127.2 | NP_780336.1 | 2, 3 | 402 | ||
| GenBank mRNA | AK076211.1 | - | 2, 3 | 408 | ||
| AK077653.1 | - | 2, 3 | 415 | |||
| AK122295.1 | - | 2, 3 | 387 | |||
| AK162091.1 | - | 2, 3 | 371 | |||
| AK162188.1 | - | 2, 3 | 283 | |||
| BC055458.1 | - | 1, 2 | 104 | |||
| BC094542.1 | - | 2, 3 | 402 | 6408744 | ||

Fbxo28
Genomic Map