siRNA Details
| Gene Symbol: Ankrd46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175134.4 | NP_780343.1 | 4 | 614 | ||
| GenBank mRNA | AB076382.1 | - | 4 | 609 | ||
| AK079270.1 | - | 2 | 173 | |||
| AK088304.1 | - | 4 | 575 | |||
| AK148049.1 | - | 4 | 575 | |||
| BC035553.1 | - | 4 | 2192 | |||
| BC086659.1 | - | 4 | 581 | 30607272 | ||
| BC094595.1 | - | 4 | 577 | 5707829 | ||
| BQ715954.1 | - | 4 | 614 | |||

Ankrd46
Genomic Map