siRNA Details
Gene Symbol: Mrps27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173757.3 | NP_776118.2 | 2, 3 | 160 | ||
GenBank mRNA | AK076008.1 | - | 2, 3 | 159 | ||
BC049149.1 | - | 2, 3 | 88 | 6309524 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mrps27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173757.3 | NP_776118.2 | 2, 3 | 160 | ||
GenBank mRNA | AK076008.1 | - | 2, 3 | 159 | ||
BC049149.1 | - | 2, 3 | 88 | 6309524 |