siRNA Details
| Gene Symbol: Ythdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173761.3 | NP_776122.1 | 5 | 2573 | ||
| GenBank mRNA | AK046768.1 | - | 5 | 2587 | ||
| AK152264.1 | - | 2 | 939 | |||
| AK168430.1 | - | 2 | 1183 | |||
| BC061479.1 | - | 5 | 2544 | 6334535 | ||
| BC065050.1 | - | 5 | 2573 | 6418891 | ||

Ythdf1
Genomic Map