siRNA Details
| Gene Symbol: Sowahd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173779.3 | NP_776140.1 | 1 | 1320 | ||
| GenBank mRNA | AK041485.1 | - | 1 | 1321 | ||
| BC107372.1 | - | 1 | 1318 | 40056413 | ||
| BC107373.1 | - | 1 | 1319 | 40056414 | ||

Sowahd
Genomic Map