siRNA Details
| Gene Symbol: C1s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173864.2 | NP_776289.2 | 11 | 1928 | ||
| GenBank mRNA | AF459020.1 | - | 11 | 1928 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1s2| Gene Symbol: C1s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173864.2 | NP_776289.2 | 11 | 1928 | ||
| GenBank mRNA | AF459020.1 | - | 11 | 1928 | ||