siRNA Details
| Gene Symbol: Ifnlr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174851.3 | NP_777276.3 | 7 | 978 | ||
| GenBank mRNA | AK141338.1 | - | 7 | 979 | ||
| AY184376.1 | - | 7 | 893 | |||
| BC057856.1 | - | 7 | 911 | 5036481 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifnlr1| Gene Symbol: Ifnlr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174851.3 | NP_777276.3 | 7 | 978 | ||
| GenBank mRNA | AK141338.1 | - | 7 | 979 | ||
| AY184376.1 | - | 7 | 893 | |||
| BC057856.1 | - | 7 | 911 | 5036481 | ||