siRNA Details
Gene Symbol: Gimap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174960.2 | NP_777620.1 | 2 | 1227 | ||
GenBank mRNA | AV291572.2 | - | 1 | 475 | ||
BC096680.1 | - | 2 | 1211 | 1345477 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gimap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174960.2 | NP_777620.1 | 2 | 1227 | ||
GenBank mRNA | AV291572.2 | - | 1 | 475 | ||
BC096680.1 | - | 2 | 1211 | 1345477 |