siRNA Details
| Gene Symbol: 4933427D06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175017.3 | NP_778182.1 | 4 | 2306 | ||
| GenBank mRNA | AK030235.1 | - | 4 | 2244 | ||
| CJ202369.1 | - | 1 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933427D06Rik| Gene Symbol: 4933427D06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175017.3 | NP_778182.1 | 4 | 2306 | ||
| GenBank mRNA | AK030235.1 | - | 4 | 2244 | ||
| CJ202369.1 | - | 1 | 90 | |||