siRNA Details
| Gene Symbol: Tmem177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175106.3 | NP_780315.1 | 2 | 1053 | ||
| GenBank mRNA | AK009043.1 | - | 2 | 1058 | ||
| AK076108.1 | - | 2 | 1059 | |||
| BC094344.1 | - | 2 | 1053 | 6402386 | ||
| BC131993.1 | - | 1 | 895 | 40130270 | ||
| BC132021.1 | - | 1 | 895 | 40130298 | ||

Tmem177
Genomic Map