siRNA Details
| Gene Symbol: Trak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041436.1 | - | 14 | 3348 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trak1| Gene Symbol: Trak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041436.1 | - | 14 | 3348 | ||