siRNA Details
| Gene Symbol: 4930430F08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175128.2 | NP_780337.2 | 7 | 1752 | ||
| GenBank mRNA | AK032226.1 | - | 7 | 1708 | ||
| AK036647.1 | - | 7 | 1735 | |||
| AK160693.1 | - | 7 | 1737 | |||
| BC082587.1 | - | 7 | 1702 | 30630155 | ||

4930430F08Rik
Genomic Map