siRNA Details
| Gene Symbol: Thap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175152.4 | NP_780361.2 | 3 | 346 | ||
| GenBank mRNA | AK008225.1 | - | 3 | 318 | ||
| AK038184.1 | - | 3 | 320 | |||
| AK050740.1 | - | 3 | 360 | |||
| BC094395.1 | - | 3 | 327 | 5700290 | ||
| BC115485.1 | - | 3 | 344 | |||
| BC115486.1 | - | 3 | 344 | |||
| BY013572.1 | - | 3 | 346 | |||

Thap3
Genomic Map