siRNA Details
| Gene Symbol: Aifm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291070.1 | NP_001277999.1 | 3 | 436 | ||
| NM_175178.4 | NP_780387.2 | 5 | 842 | |||
| GenBank mRNA | AK012958.1 | - | 2 | 103 | ||
| AK049928.1 | - | 5 | 673 | |||
| AK158809.1 | - | 4 | 504 | |||
| BC046517.1 | - | 5 | 828 | |||
| BC096476.1 | - | 4 | 617 | 4221224 | ||
| BC120685.1 | - | 4 | 471 | 40129608 | ||

Aifm3
Genomic Map