siRNA Details
| Gene Symbol: Prr5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083810.2 | NP_001077279.1 | 9 | 1151 | ||
| NM_001110849.1 | NP_001104319.1 | 10 | 1014 | |||
| NM_175181.5 | NP_780390.3 | 9 | 1100 | |||
| GenBank mRNA | AK011178.1 | - | 4 | 348 | ||
| AK030499.1 | - | 9 | 1058 | |||
| AK145981.1 | - | 9 | 1102 | |||
| BC048394.1 | - | 9 | 1010 | 6391924 | ||
| BC172132.1 | - | 8 | 826 | 9054802 | ||
| BC172133.1 | - | 8 | 822 | |||
| BC172158.1 | - | 7 | 734 | |||

Prr5l
Genomic Map