siRNA Details
| Gene Symbol: Mvb12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175184.4 | NP_780393.2 | 3 | 800 | ||
| GenBank mRNA | AK012178.1 | - | 3 | 293 | ||
| AK087821.1 | - | 3 | 318 | |||
| AK131113.1 | - | 3 | 318 | |||
| AK154267.1 | - | 3 | 294 | |||
| BC049129.1 | - | 3 | 313 | 6825067 | ||
| BC059907.1 | - | 3 | 800 | 6824889 | ||

Mvb12b
Genomic Map