siRNA Details
| Gene Symbol: Prox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175198.4 | NP_780407.1 | 2 | 576 | ||
| GenBank mRNA | AK006828.1 | - | 1 | 442 | ||
| AK029978.1 | - | 2 | 532 | |||
| AK051700.1 | - | 2 | 577 | |||
| BC119330.1 | - | 1 | 320 | 8734079 | ||
| BC119332.1 | - | 1 | 320 | 8734081 | ||

Prox2
Genomic Map