siRNA Details
| Gene Symbol: Hspa12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175199.3 | NP_780408.1 | 2 | 200 | ||
| GenBank mRNA | AB093239.1 | - | 2 | 144 | ||
| AK006867.1 | - | 2 | 184 | |||
| AK038491.1 | - | 2 | 172 | |||
| AK087732.1 | - | 3 | 356 | |||
| AK141925.1 | - | 2 | 201 | |||
| AK165073.1 | - | 2 | 228 | |||
| AY196789.1 | - | 2 | 199 | |||
| BC030362.1 | - | 2 | 170 | 5369051 | ||

Hspa12a
Genomic Map