siRNA Details
| Gene Symbol: 4930544M13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076988.1 | - | 2 | 492 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_175218.1 | - | 492 | Chr. - - [-] on Build | ||

4930544M13Rik
Genomic Map