siRNA Details
| Gene Symbol: 2310022B05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175149.4 | NP_780358.3 | 1, 2 | 530 | ||
| GenBank mRNA | AK009458.1 | - | 1, 2 | 355 | ||
| AK084773.1 | - | 1, 2 | 349 | |||
| AK154813.1 | - | 1, 2 | 356 | |||
| AK155222.1 | - | 1, 2 | 385 | |||
| AK170562.1 | - | 1, 2 | 375 | |||
| BC058626.1 | - | 1, 2 | 530 | 6401577 | ||

2310022B05Rik
Genomic Map