siRNA Details
| Gene Symbol: Galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291002.1 | NP_001277931.1 | 4, 5 | 466 | ||
| NM_175154.3 | NP_780363.1 | 4, 5 | 572 | |||
| GenBank mRNA | AK053002.1 | - | 4, 5 | 422 | ||
| AK082521.1 | - | 4, 5 | 463 | |||
| AK152851.1 | - | 2, 3 | 155 | |||
| AK154762.1 | - | 4, 5 | 421 | |||
| AK171939.1 | - | 4, 5 | 449 | |||
| BC079843.1 | - | 4, 5 | 436 | 5690056 | ||

Galk2
Genomic Map