siRNA Details
| Gene Symbol: 5430419D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175166.3 | NP_780375.2 | 3, 4 | 215 | ||
| GenBank mRNA | AK035662.1 | - | 3, 4 | 216 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5430419D17Rik| Gene Symbol: 5430419D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175166.3 | NP_780375.2 | 3, 4 | 215 | ||
| GenBank mRNA | AK035662.1 | - | 3, 4 | 216 | ||