siRNA Details
| Gene Symbol: Plekhf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175175.4 | NP_780384.1 | 2 | 1000 | ||
| GenBank mRNA | AK033666.1 | - | 2 | 994 | ||
| AK154634.1 | - | 2 | 991 | |||
| BC016134.1 | - | 2 | 941 | 4503757 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhf2| Gene Symbol: Plekhf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175175.4 | NP_780384.1 | 2 | 1000 | ||
| GenBank mRNA | AK033666.1 | - | 2 | 994 | ||
| AK154634.1 | - | 2 | 991 | |||
| BC016134.1 | - | 2 | 941 | 4503757 | ||