siRNA Details
| Gene Symbol: Erich3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175176.3 | NP_780385.1 | 2 | 330 | ||
| GenBank mRNA | AK076632.1 | - | 2 | 331 | ||
| BC138236.1 | - | 2 | 250 | 8861255 | ||
| BC145840.1 | - | 2 | 250 | 40131143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich3| Gene Symbol: Erich3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175176.3 | NP_780385.1 | 2 | 330 | ||
| GenBank mRNA | AK076632.1 | - | 2 | 331 | ||
| BC138236.1 | - | 2 | 250 | 8861255 | ||
| BC145840.1 | - | 2 | 250 | 40131143 | ||