siRNA Details
| Gene Symbol: Hepacam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175189.4 | NP_780398.2 | 5 | 897 | ||
| GenBank mRNA | AK013637.1 | - | 5 | 897 | ||
| AK081154.1 | - | 5 | 813 | |||
| AK138438.1 | - | 4 | 421 | |||
| AK158265.1 | - | 5 | 1135 | |||
| BC082537.1 | - | 5 | 811 | |||
| BC139057.1 | - | 5 | 887 | 8862079 | ||
| BC139058.1 | - | 5 | 887 | 8862080 | ||

Hepacam
Genomic Map