siRNA Details
| Gene Symbol: AP1G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030007.1 | NP_001025178.1 | 24 | 2928 | ||
| NM_001128.5 | NP_001119.3 | 23 | 2919 | |||
| GenBank mRNA | AB015317.1 | - | 23 | 2624 | ||
| AK055122.1 | - | 7 | 1539 | |||
| AK128078.1 | - | 26 | 3474 | |||
| AK293233.1 | - | 15 | 1543 | |||
| AL110198.1 | - | 7 | 993 | |||
| BC036283.1 | - | 24 | 2893 | 4158263 | ||

AP1G1
Genomic Map