siRNA Details
| Gene Symbol: CRISP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131.2 | NP_001122.2 | 8 | 1782 | ||
| NM_001205220.1 | NP_001192149.1 | 8 | 1793 | |||
| NM_170609.1 | NP_733758.1 | 7 | 1693 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRISP1| Gene Symbol: CRISP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131.2 | NP_001122.2 | 8 | 1782 | ||
| NM_001205220.1 | NP_001192149.1 | 8 | 1793 | |||
| NM_170609.1 | NP_733758.1 | 7 | 1693 | |||