siRNA Details
| Gene Symbol: Rif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175238.5 | NP_780447.4 | 30 | 5853 | ||
| GenBank mRNA | AK030007.1 | - | 1 | 1362 | ||
| AK083291.1 | - | 1 | 355 | |||
| AK088917.1 | - | 1 | 887 | |||
| AY428571.2 | - | 30 | 5837 | |||
| AY585206.1 | - | 29 | 5846 | |||
| BC055320.1 | - | 1 | 213 | |||
| JX255737.1 | - | 29 | 5789 | |||

Rif1
Genomic Map