siRNA Details
| Gene Symbol: Tmem127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175145.3 | NP_780354.1 | 4 | 3712 | ||
| GenBank mRNA | AK009130.1 | - | 1 | 247 | ||
| AK033561.1 | - | 1 | 1425 | |||
| AK165880.1 | - | 4 | 3780 | |||
| BC022794.1 | - | 1 | 2851 | |||
| BC049832.1 | - | 4 | 3714 | 6313802 | ||

Tmem127
Genomic Map