siRNA Details
| Gene Symbol: Tsr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164578.1 | NP_001158050.1 | 5 | 1721 | ||
| NM_175146.4 | NP_780355.2 | 5 | 1712 | |||
| GenBank mRNA | AK044504.1 | - | 5 | 1687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsr2| Gene Symbol: Tsr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164578.1 | NP_001158050.1 | 5 | 1721 | ||
| NM_175146.4 | NP_780355.2 | 5 | 1712 | |||
| GenBank mRNA | AK044504.1 | - | 5 | 1687 | ||