siRNA Details
| Gene Symbol: Thap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145929.1 | NP_001139401.1 | 3 | 464 | ||
| NM_175152.4 | NP_780361.2 | 4 | 566 | |||
| GenBank mRNA | AK008225.1 | - | 4 | 538 | ||
| AK038184.1 | - | 4 | 540 | |||
| AK050740.1 | - | 4 | 580 | |||
| BC059091.1 | - | 3 | 449 | |||
| BC094395.1 | - | 4 | 547 | 5700290 | ||
| BC115485.1 | - | 4 | 565 | |||
| BC115486.1 | - | 4 | 563 | |||

Thap3
Genomic Map