siRNA Details
Gene Symbol: Zfp689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175163.3 | NP_780372.2 | 3 | 2284 | ||
GenBank mRNA | AK028417.1 | - | 3 | 2248 | ||
AK051667.1 | - | 3 | 2098 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175163.3 | NP_780372.2 | 3 | 2284 | ||
GenBank mRNA | AK028417.1 | - | 3 | 2248 | ||
AK051667.1 | - | 3 | 2098 |